Merck
Regulation of MEIS1 by distal enhancer elements in acute leukemia.
Leukemia (2013-09-12)
Q F Wang, Y J Li, J F Dong, B Li, J J Kaberlein, L Zhang, F E Arimura, R T Luo, J Ni, F He, J Wu, R Mattison, J Zhou, C Z Wang, S Prabhakar, M A Nobrega, M J Thirman
ABSTRACT
Aberrant activation of the three-amino-acid-loop extension homeobox gene MEIS1 shortens the latency and accelerates the onset and progression of acute leukemia, yet the molecular mechanism underlying persistent activation of the MEIS1 gene in leukemia remains poorly understood. Here we used a combined comparative genomics analysis and an in vivo transgenic zebrafish assay to identify six regulatory DNA elements that are able to direct green fluorescent protein expression in a spatiotemporal manner during zebrafish embryonic hematopoiesis. Analysis of chromatin characteristics and regulatory signatures suggests that many of these predicted elements are potential enhancers in mammalian hematopoiesis. Strikingly, one of the enhancer elements (E9) is a frequent integration site in retroviral-induced mouse acute leukemia. The genomic region corresponding to enhancer E9 is differentially marked by H3K4 monomethylation and H3K27 acetylation, hallmarks of active enhancers, in multiple leukemia cell lines. Decreased enrichment of these histone marks is associated with downregulation of MEIS1 expression during hematopoietic differentiation. Further, MEIS1/HOXA9 transactivate this enhancer via a conserved binding motif in vitro, and participate in an autoregulatory loop that modulates MEIS1 expression in vivo. Our results suggest that an intronic enhancer regulates the expression of MEIS1 in hematopoiesis and contributes to its aberrant expression in acute leukemia.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Hox A9 Antibody, Upstate®, from rabbit