Merck
  • Eicosapentaenoic acid (EPA) has optimal chain length and degree of unsaturation to inhibit oxidation of small dense LDL and membrane cholesterol domains as compared to related fatty acids in vitro.
Eicosapentaenoic acid (EPA) has optimal chain length and degree of unsaturation to inhibit oxidation of small dense LDL and membrane cholesterol domains as compared to related fatty acids in vitro.
Biochimica et biophysica acta. Biomembranes (2020-03-07)
Samuel C R Sherratt, Rebecca A Juliano, R Preston Mason
ABSTRACT
Oxidation of small dense low-density lipoprotein (sdLDL) and membranes is causally related to atherosclerosis. The omega-3 fatty acid (FA) eicosapentaenoic acid (EPA, 20:5, ω-3) significantly reduced oxidized LDL in patients with hypertriglyceridemia by unknown mechanisms. We compared EPA effects to related FAs of varying chain length and unsaturation on oxidation of sdLDL and model membranes, and on cholesterol crystal domains. We compared EPA to the FAs: stearic (SA, 18:0), oleic (OA, 18:1, ω-9), linoleic (LA, 18:2, ω-6), alpha-linolenic (ALA, 18:3, ω-3), eicosanoic (EA, 20:0), eicosatrienoic (ETE, 20:3, ω-3), arachidonic (AA, 20:4, ω-6), docosapentaenoic (DPA, 22:5, ω-3), and docosahexaenoic (DHA, 22:6, ω-3). Human sdLDL or model membranes of cholesterol and 1,2-Dilinoleoyl-sn-glycero-3-phosphocholine [18:2(cis)PC or DLPC] were preincubated with FAs followed by copper-induced oxidation. Malondialdehyde (MDA) or lipid hydroperoxides (LOOH) levels measured oxidation; small-angle X-ray diffraction assessed cholesterol domain formation. After 40 min, EPA reduced MDA levels 70% compared to vehicle (p < 0.001). Lesser inhibition was observed with DHA, DPA, ETE, and ALA (33%, 34%, 32%, and 16%, respectively; all p < 0.001 versus vehicle). Similar relative FA effects were observed in model membranes where EPA more substantially inhibited cholesterol crystal domain formation. We observed relationships between hydrocarbon length and unsaturation with antioxidant activity and membrane cholesterol domain formation. EPA had the most favorable molecular structure, likely contributing to membrane stability, improved lipoprotein clearance, and reduced inflammation. Insight is provided into FA hydrocarbon length and unsaturation relationships with antioxidant activity in lipoproteins and membranes, and cholesterol crystal domains formation.
Sigma-Aldrich
2-(Diisopropylamino)ethyl methacrylate, 97%, contains ~100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
1,2-Dilinoleoyl-sn-glycero-3-phosphocholine, ≥99% (TLC), lyophilized powder
Avanti
18:2 (Cis) PC (DLPC), 1,2-dilinoleoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
18:2 (Cis) PC (DLPC), Avanti Research - A Croda Brand