Merck
  • Discovery of gene regulatory elements through a new bioinformatics analysis of haploid genetic screens.
Discovery of gene regulatory elements through a new bioinformatics analysis of haploid genetic screens.
PloS one (2019-01-30)
Bhaven B Patel, Andres M Lebensohn, Ganesh V Pusapati, Jan E Carette, Julia Salzman, Rajat Rohatgi
ABSTRACT
The systematic identification of regulatory elements that control gene expression remains a challenge. Genetic screens that use untargeted mutagenesis have the potential to identify protein-coding genes, non-coding RNAs and regulatory elements, but their analysis has mainly focused on identifying the former two. To identify regulatory elements, we conducted a new bioinformatics analysis of insertional mutagenesis screens interrogating WNT signaling in haploid human cells. We searched for specific patterns of retroviral gene trap integrations (used as mutagens in haploid screens) in short genomic intervals overlapping with introns and regions upstream of genes. We uncovered atypical patterns of gene trap insertions that were not predicted to disrupt coding sequences, but caused changes in the expression of two key regulators of WNT signaling, suggesting the presence of cis-regulatory elements. Our methodology extends the scope of haploid genetic screens by enabling the identification of regulatory elements that control gene expression.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Tablets, For General Use
Sigma-Aldrich
Anti-LRP6, clone A59, Ectodomain Antibody, clone A59, from mouse