Skip to Content
Merck
Triphenylsilanethiol 98%

Triphenylsilanethiol

Triphenylsilanethiol 98%
Synonym(s):
Mercaptotriphenylsilane
Linear Formula:
(C6H5)3SiSH
CAS Number:
Molecular Weight:
292.47

Products

Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing