Skip to Content
Merck
Triphenylene BCR®, certified reference material

Triphenylene

Triphenylene BCR®, certified reference material
Synonym(s):
9,10-Benzophenanthrene
Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing