
Trilaurin

Synonym(s):
Glyceryl tridodecanoate, 1,2,3-Tridodecanoylglycerol, 1,2,3-Trilauroylglycerol, Glycerol trilaurate, Glyceryl trilaurate, Tridodecanoin, Trilaurin
Linear Formula:
[CH3(CH2)10COOCH2]2CHOCO(CH2)10CH3
CAS Number:
Molecular Weight:
639.00
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing