
Sodium octanoate-1-13C

Synonym(s):
13C Labeled sodium octanoate, Caprylic acid-1-13C sodium salt, Octanoic acid-1-13C sodium salt, Sodium caprylate-1-13C
Linear Formula:
CH3(CH2)613CO2Na
CAS Number:
Molecular Weight:
167.19
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing