
Rosuvastatin Ketone

Synonym(s):
(3R,6E)-7-[4-(4-Fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]pyrimidin-5-yl]-3-hydroxy-5-oxohept-6-enoic acid, (3R,6E)-7-[4-(4-Fluorophenyl)-2-(N-methylmethanesulfonamido)-6-(propan-2-yl)pyrimidin-5-yl]-3-hydroxy-5-oxohept-6-enoic acid, 5-Oxorosuvastatin
Empirical Formula (Hill Notation):
C22H26FN3O6S
CAS Number:
Molecular Weight:
479.52
Products
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing