
Risedronate impurity E

Synonym(s):
2-(3-Pyridinyl)ethylidene]bis(phosphonic acid), 2-(Pyridin-3-yl)ethane-1,1-diyl]bis(phosphonic acid)
Empirical Formula (Hill Notation):
C7H11NO6P2
CAS Number:
Molecular Weight:
267.11
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing