
Pseudohypericin

Empirical Formula (Hill Notation):
C30H16O9
CAS Number:
Molecular Weight:
520.44
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing