
Phenobarbital-D5 (side chain) solution

Empirical Formula (Hill Notation):
C12D5H7N2O3
CAS Number:
Molecular Weight:
237.27
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing