
Pentafluorophenylmagnesium bromide solution

Linear Formula:
C6F5MgBr
CAS Number:
Molecular Weight:
271.27
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing