
Penciclovir

Synonym(s):
2-Amino-1,9-dihydro-9-[4-hydroxy-3-(hydroxymethyl)butyl]-6H-purin-6-one, BRL-39123
Empirical Formula (Hill Notation):
C10H15N5O3
CAS Number:
Molecular Weight:
253.26
Products
Product Category
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing