
Neosaxitoxin solution

Synonym(s):
neo-Saxitoxin solution, NeoSTX
Empirical Formula (Hill Notation):
C10H17N7O5
CAS Number:
Molecular Weight:
315.29
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1