
Myoglobin from equine skeletal muscle

Synonym(s):
Myoglobin from horse skeletal muscle
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Physical Form
Purity
Tissue
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing