
Methylprednisolone hemisuccinate

Synonym(s):
11β,17,21-Trihydroxy-6α-methylpregna-1,4-diene-3,20-dione 21-succinate, Methylprednisolone hydrogen succinate
Empirical Formula (Hill Notation):
C26H34O8
CAS Number:
Molecular Weight:
474.54
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing