Skip to Content
Merck
Menthone analytical standard

Menthone

Menthone analytical standard
Synonym(s):
2-Isopropyl-5-methylcyclohexanone
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing