
Lisinopril Impurity B

Synonym(s):
p-Toluenesulfonic acid monohydrate
Linear Formula:
CH3C6H4SO3H · H2O
CAS Number:
Molecular Weight:
190.22
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing