
Isoginkgetin

Synonym(s):
Pre-mRNA Splicing Inhibitor, Isoginkgetin, 4′,4′′′-Dimethylamentoflavone, 4′,4′′′-dimethylamentoflavone;5,5′′,7,7′′-tetrahydroxy-4′,4′′′-dimethoxy-3′′′,8-biflavone, Amentoflavone 4′,4′′′-dimethyl ether
Empirical Formula (Hill Notation):
C32H22O10
CAS Number:
Molecular Weight:
566.51
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing