
Ibuprofen Impurity K

Synonym(s):
(2RS)-2-(4-Formylphenyl)propionic acid, (2RS)-2-(4-Formylphenyl)propanoic acid
Empirical Formula (Hill Notation):
C10H10O3
CAS Number:
Molecular Weight:
178.18
Products
Brand
Formula Weight
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing