
Glycyl-L-tyrosine

Synonym(s):
Glycyl-L-tyrosine dihydrate
Empirical Formula (Hill Notation):
C11H14N2O4 · 2H2O
CAS Number:
Molecular Weight:
274.27
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing