
Fusicoccin from Fusicoccum amygdali

Empirical Formula (Hill Notation):
C36H56O12
CAS Number:
Molecular Weight:
680.82
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing