
Furosemide impurity A

Synonym(s):
2-Chloro-4-furfurylamino-5-sulfamoylbenzoic acid
Empirical Formula (Hill Notation):
C12H11ClN2O5S
CAS Number:
Molecular Weight:
330.74
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing