
Fluconazole Related Compound A

Synonym(s):
2-[2-Fluoro-4-(1H-1,2,4-triazol-1-yl)phenyl]-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol
Empirical Formula (Hill Notation):
C15H14FN9O
CAS Number:
Molecular Weight:
355.33
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2