
Exenatide

Synonym(s):
Exendin-4
Empirical Formula (Hill Notation):
C184H282N50O60S
CAS Number:
Molecular Weight:
4186.57
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing