
di-Potassium oxalate monohydrate

Synonym(s):
di-Potassium oxalate monohydrate
Empirical Formula (Hill Notation):
C2K2O4 · xH2O
CAS Number:
Molecular Weight:
166.22 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing