Skip to Content
Merck
Diisopropylamine, polymer-bound 100-200 mesh, extent of labeling: 2.0-3.5 mmol/g loading, 1 % cross-linked with divinylbenzene

Diisopropylamine, polymer-bound

Diisopropylamine, polymer-bound 100-200 mesh, extent of labeling: 2.0-3.5 mmol/g loading, 1 % cross-linked with divinylbenzene
Synonym(s):
Benzyldiisopropylamine, polymer-bound, Copolymer of styrene and divinylbenzene, diisopropylaminomethylated, Diisopropylaminomethyl-polystyrene, Polystyrene cross-linked with divinylbenzene, diisopropylaminomethylated, ‘Hünig’s base’ on polystyrene

Products

Brand
Chain Type
Reaction Type
Reactivity