
Dicyanocobinamide

Synonym(s):
Cobinamide dicyanide, Etiocobalamin, Factor B
Empirical Formula (Hill Notation):
C50H72CoN13O8
CAS Number:
Molecular Weight:
1042.12
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing