
Demeclocycline for system suitability

Synonym(s):
Demeclocycline hydrochloride, 7-Chloro-6-demethyltetracycline hydrochloride, Demethylchlorotetracycline hydrochloride
Empirical Formula (Hill Notation):
C21H21ClN2O8 · HCl
CAS Number:
Molecular Weight:
501.31
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing