
(+)-Cinchonine

Synonym(s):
Cinchonine monohydrochloride dihydrate, NSC 6176, (+)-Cinchonine, Cinchonine
Empirical Formula (Hill Notation):
C19H22N2O
CAS Number:
Molecular Weight:
294.39
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing