
Cytidine

Synonym(s):
Cytosine β-D-riboside, Cytosine-1-β-D-ribofuranoside, Cytidine
Empirical Formula (Hill Notation):
C9H13N3O5
CAS Number:
Molecular Weight:
243.22
Products
Brand
Biological Source
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Special Grade
Technique
Showing 1-3 of 3