
Clobetasol Propionate Related Compound A

Synonym(s):
(17R)-4′-Chloro-5′-ethyl-9-fluoro-11β-hydroxy-16β-methylspiro[androsta-1,4-diene-17,2′(3′H)-furan]-3,3′-dione
Empirical Formula (Hill Notation):
C25H30ClFO4
CAS Number:
Molecular Weight:
448.95
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing