
cis-3-Hexenyl hexanoate

Linear Formula:
CH3(CH2)4CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
198.30
Products
Brand
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing