
Ciprofloxacin formamide

Synonym(s):
1-Cyclopropyl-6-fluoro-7-(4-formyl-1-piperazinyl)-1,4-dihydro-4-oxo-3-quinolonecarboxylic acid, N-Formylciprofloxacin
Empirical Formula (Hill Notation):
C18H18FN3O4
CAS Number:
Molecular Weight:
359.35
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2