
Cefepime hydrochloride

Synonym(s):
Cefepime dihydrochloride monohydrate, Cefepime hydrochloride
Linear Formula:
C19H25N6O5S2Cl · HCl · H2O
CAS Number:
Molecular Weight:
571.50
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3