
Carvedilol for system suitability

Synonym(s):
Carvedilol, 1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-2-propanol, BM-14190
Empirical Formula (Hill Notation):
C24H26N2O4
CAS Number:
Molecular Weight:
406.47
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing