
Carfentrazone-ethyl

Synonym(s):
Ethyl 2-chloro-3-{2-chloro-5-[4-(difluoromethyl)-4,5-dihydro-3-methyl-5-oxo-1H-1,2,4-triazol-1-yl]-4-fluoro-phenyl}propionate, Ethyl-α,2-dichloro-5-[4-(difluoromethyl)-4,5-dihydro-3-methyl-5-oxo-1H-1,2,4-triazol-1-yl]-4-fluorobenzenepropanoate
Empirical Formula (Hill Notation):
C15H14Cl2F3N3O3
CAS Number:
Molecular Weight:
412.19
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2