
Bromomethane solution

Synonym(s):
Methyl bromide
Empirical Formula (Hill Notation):
CH3Br
CAS Number:
Molecular Weight:
94.94
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing