
Brazilin

Synonym(s):
Brasilin, 6,6a,7,11b-Tetrahydro-3,6a,9,10-tetrahydroxyindeno[2,1-c]chromene, Natural Red 24
Empirical Formula (Hill Notation):
C16H14O5
CAS Number:
Molecular Weight:
286.28
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Product Line
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing