
Benzhydrol

Synonym(s):
Diphenylmethanol, Benzhydrol, Benzhydryl alcohol, Diphenyl carbinol
Linear Formula:
(C6H5)2CHOH
CAS Number:
Molecular Weight:
184.23
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing