
β-(1,2,4-Triazol-3-yl)-DL-alanine

Synonym(s):
(±)-2-Amino-3-(1,2,4-triazol-3-yl)propionic acid, DL-1,2,4-Triazole-3-alanine
Empirical Formula (Hill Notation):
C5H8N4O2
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing