
Anti-TRAF2 antibody produced in rabbit

Synonym(s):
Anti-TNF receptor-associated factor 2 antibody produced in rabbit, Anti-Tumor necrosis factor type 2 receptor-associated protein 3 antibody produced in rabbit, Anti-TNF receptor-associated factor 2
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing