
Amikacin

Synonym(s):
Amikacin
Empirical Formula (Hill Notation):
C22H43N5O13
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3