
6-Methylchrysene

Empirical Formula (Hill Notation):
C19H14
CAS Number:
Molecular Weight:
242.31
Products
Formula Weight
Manufacturer
Agency
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing