
5α-Pregnan-3α-ol-20-one

Synonym(s):
3α-Hydroxy-5α-pregnan-20-one, 3α-OH-DHP, Allopregnan-3α-ol-20-one, Allopregnanolone, Brexanolone
Empirical Formula (Hill Notation):
C21H34O2
CAS Number:
Molecular Weight:
318.49
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing