
4-Methylaminoantipyrine hydrochloride

Synonym(s):
1,2-Dihydro-1,5-dimethyl-4-(methylamino)-2-phenyl-3H-pyrazol-3-one monohydrochloride, N-Methylaminophenazone, Noramidopyrine hydrochloride, Noraminopyrine hydrochloride
Empirical Formula (Hill Notation):
C12H15N3O · HCl
CAS Number:
Molecular Weight:
253.73
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing