
3-Isobutyl-1-methylxanthine

Synonym(s):
1-Methyl-3-isobutylxanthine, 3,7-Dihydro-1-methyl-3-(2-methylpropyl)-1H-purine-2,6-dione, 3-Isobutyl-1-methyl-2,6(1H,3H)-purinedione, IBMX, 3-Isobutyl-1-methylxanthine
Empirical Formula (Hill Notation):
C10H14N4O2
CAS Number:
Molecular Weight:
222.24
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Showing 1-3 of 3