
3,3′-Dipropylthiacarbocyanine iodide

Synonym(s):
3-Propyl-2-[3-[3-propyl-2(3H)benzothiazolylidene]-1-propenyl]benzothiazolium iodide, DiSC3(3)
Empirical Formula (Hill Notation):
C23H25IN2S2
CAS Number:
Molecular Weight:
520.49
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing