Skip to Content
Merck
2-Furoic acid analytical standard

2-Furoic acid

2-Furoic acid analytical standard
Synonym(s):
Furan-2-carboxylic acid
Empirical Formula (Hill Notation):
C5H4O3
CAS Number:
Molecular Weight:
112.08
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing