Skip to Content
Merck
2-(2-Methoxyethoxy)acetic acid technical grade

2-(2-Methoxyethoxy)acetic acid

2-(2-Methoxyethoxy)acetic acid technical grade
Synonym(s):
O-(2-Methoxyethyl)glycolic acid
Linear Formula:
CH3OCH2CH2OCH2CO2H
CAS Number:
Molecular Weight:
134.13
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Monomer Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing